accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7318-00-5

IUPAC Name: ethyl (2E)-3-aminobut-2-enoate
Synonyms: ethyl (2E)-3-aminobut-2-enoate
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: YPMPTULBFPFSEQ-SNAWJCMRSA-N
SMILES: CCOC(=O)\C=C(/C)N
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)