accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOBr

CAS RN 7321-58-6

IUPAC Name: 1-bromo-2-methanesulfinylbenzene
Synonyms: 1-bromo-2-methanesulfinylbenzene
Molecular Weight (g/mol): 219.1
Molecular Formula: C7H7BrOS
InChi Key: NNNHMHLNXXGWBA-UHFFFAOYNA-N
SMILES: CS(=O)C1=CC=CC=C1Br
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results