accessibility menu, dialog, popup

UserName

Viewing profile as user:
FIFFFFSOOO

CAS RN 732306-64-8

IUPAC Name: bis(4-fluorophenyl)iodanium trifluoromethanesulfonate
Synonyms: bis(4-fluorophenyl)iodanium triflate
Molecular Weight (g/mol): 466.16
Molecular Formula: C13H8F5IO3S
InChi Key: HTFNIUDXMGKTQK-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.FC1=CC=C([I+]C2=CC=C(F)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results