accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73278-90-7

IUPAC Name: methyl 3-[(piperidin-1-yl)methyl]benzoate
Synonyms: methyl 3-(piperidin-1-ylmethyl)benzoate
Molecular Weight (g/mol): 233.31
Molecular Formula: C14H19NO2
InChi Key: RJOMIJDQGNNOCO-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(CN2CCCCC2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results