accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73318-02-2

NNNNFF

CAS RN 73318-02-2

IUPAC Name: 2-[4-(dicyanomethylidene)-2,5-difluorocyclohexa-2,5-dien-1-ylidene]propanedinitrile
Synonyms: 2-[4-(dicyanomethylidene)-2,5-difluorocyclohexa-2,5-dien-1-ylidene]propanedinitrile
Molecular Weight (g/mol): 240.17
Molecular Formula: C12H2F2N4
InChi Key: PFSDYUVXXQTNMX-UHFFFAOYSA-N
SMILES: FC1=CC(=C(C#N)C#N)C(F)=CC1=C(C#N)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results