accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONOOH3C

CAS RN 73323-65-6

IUPAC Name: 1-tert-butyl 2-methyl pyrrolidine-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-methyl pyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 229.28
Molecular Formula: C11H19NO4
InChi Key: WVDGSSCWFMSRHN-UHFFFAOYNA-N
SMILES: COC(=O)C1CCCN1C(=O)OC(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results