accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOSS

CAS RN 7333-07-5

IUPAC Name: bis(thiophen-2-yl)ethane-1,2-dione
Synonyms: bis(thiophen-2-yl)ethane-1,2-dione
Molecular Weight (g/mol): 222.28
Molecular Formula: C10H6O2S2
InChi Key: UNWKVSDABPCZMK-UHFFFAOYSA-N
SMILES: O=C(C(=O)C1=CC=CS1)C1=CC=CS1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results