accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNBr

CAS RN 73335-64-5

IUPAC Name: [(5-bromopyridin-3-yl)methyl](methyl)amine
Synonyms: [(5-bromopyridin-3-yl)methyl](methyl)amine
Molecular Weight (g/mol): 201.07
Molecular Formula: C7H9BrN2
InChi Key: UHDHQYPCHVXIKK-UHFFFAOYSA-N
SMILES: CNCC1=CC(Br)=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results