accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONHCH3H3CH2OHCl

CAS RN 73360-54-0

IUPAC Name: 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide hydrate hydrochloride
Synonyms: bupivacaine hydrate hydrochloride
Molecular Weight (g/mol): 342.91
Molecular Formula: C18H31ClN2O2
InChi Key: HUCIWBPMHXGLFM-UHFFFAOYNA-N
SMILES: O.Cl.CCCCN1CCCCC1C(=O)NC1=C(C)C=CC=C1C
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results