accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OHC

CAS RN 7341-96-0

IUPAC Name: prop-2-ynamide
Synonyms: propiolamide
Molecular Weight (g/mol): 69.06
Molecular Formula: C3H3NO
InChi Key: HCJTYESURSHXNB-UHFFFAOYSA-N
SMILES: NC(=O)C#C
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)