accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHOOHHCl

CAS RN 73415-84-6

IUPAC Name: 2-(piperidin-4-yl)acetic acid hydrochloride
Synonyms: piperidin-4-ylacetic acid hydrochloride
Molecular Weight (g/mol): 179.64
Molecular Formula: C7H14ClNO2
InChi Key: IYDUFJOXYMLYNM-UHFFFAOYSA-N
SMILES: Cl.OC(=O)CC1CCNCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)