accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7353-91-5

H3CNCH3MgBr

CAS RN 7353-91-5

IUPAC Name: 4-(bromomagnesio)-N,N-dimethylaniline
Synonyms: 4-(bromomagnesio)-N,N-dimethylaniline
Molecular Weight (g/mol): 224.38
Molecular Formula: C8H10BrMgN
InChi Key: OWWWKAXERYRWAU-UHFFFAOYSA-M
SMILES: CN(C)C1=CC=C([Mg]Br)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results