accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7355-18-2

IUPAC Name: methyl 3,4,5,6-tetrakis(acetyloxy)oxane-2-carboxylate
Synonyms: methyl 3,4,5,6-tetrakis(acetyloxy)oxane-2-carboxylate
Molecular Weight (g/mol): 376.31
Molecular Formula: C15H20O11
InChi Key: DPOQCELSZBSZGX-UHFFFAOYNA-N
SMILES: COC(=O)C1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results