accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOBrOHHO

CAS RN 7355-22-8

IUPAC Name: 5-bromo-2,4-dihydroxybenzoic acid
Synonyms: 5-bromo-2,4-dihydroxybenzoic acid
Molecular Weight (g/mol): 233.02
Molecular Formula: C7H5BrO4
InChi Key: ZRBCISXJLHZOMS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Br)=C(O)C=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results