accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7355-55-7

IUPAC Name: 2-amino-1H,4H,7H-pyrrolo[2,3-d]pyrimidin-4-one
Synonyms: 2-amino-1H,7H-pyrrolo[2,3-d]pyrimidin-4-one
Molecular Weight (g/mol): 150.14
Molecular Formula: C6H6N4O
InChi Key: OLAFFPNXVJANFR-UHFFFAOYSA-N
SMILES: NC1=NC(=O)C2=C(NC=C2)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results