accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHCl

CAS RN 7355-58-0

IUPAC Name: N-(2-chloroethyl)acetamide
Synonyms: N-(2-chloroethyl)acetamide
Molecular Weight (g/mol): 121.56
Molecular Formula: C4H8ClNO
InChi Key: HSKNJSHFPPHTAQ-UHFFFAOYSA-N
SMILES: CC(=O)NCCCl
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)