accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73579-08-5

IUPAC Name: N,1-dimethylpiperidin-4-amine
Synonyms: N,1-dimethylpiperidin-4-amine
Molecular Weight (g/mol): 128.22
Molecular Formula: C7H16N2
InChi Key: XRYGCVVVDCEPRL-UHFFFAOYSA-N
SMILES: CNC1CCN(C)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results