accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73583-39-8

IUPAC Name: 3-bromo-5-chloropyridine
Synonyms: 3-bromo-5-chloropyridine
Molecular Weight (g/mol): 192.44
Molecular Formula: C5H3BrClN
InChi Key: BELDOPUBSLPBCQ-UHFFFAOYSA-N
SMILES: ClC1=CN=CC(Br)=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)