accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOClOClOCl

CAS RN 7359-58-2

IUPAC Name: tris(4-chlorophenyl) borate
Synonyms: tris(4-chlorophenyl) borate
Molecular Weight (g/mol): 393.45
Molecular Formula: C18H12BCl3O3
InChi Key: JOAZIIBFQMIXJM-UHFFFAOYSA-N
SMILES: ClC1=CC=C(OB(OC2=CC=C(Cl)C=C2)OC2=CC=C(Cl)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results