accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73616-27-0

IUPAC Name: 2-(5-amino-1H-pyrazol-1-yl)ethan-1-ol
Synonyms: 2-(5-aminopyrazol-1-yl)ethanol
Molecular Weight (g/mol): 127.15
Molecular Formula: C5H9N3O
InChi Key: IHQRJCVJAUKIEP-UHFFFAOYSA-N
SMILES: NC1=CC=NN1CCO
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)