accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HOOOO

CAS RN 7362-93-8

IUPAC Name: 3-amino-4-(benzyloxy)-4-oxobutanoic acid
Synonyms: 3-amino-4-(benzyloxy)-4-oxobutanoic acid
Molecular Weight (g/mol): 223.23
Molecular Formula: C11H13NO4
InChi Key: NJSRYBIBUXBNSW-UHFFFAOYNA-N
SMILES: NC(CC(O)=O)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results