accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7367-88-6

IUPAC Name: ethyl (2E)-dec-2-enoate
Synonyms: ethyl (2E)-dec-2-enoate
Molecular Weight (g/mol): 198.31
Molecular Formula: C12H22O2
InChi Key: GNJARWZWODMTDR-ZHACJKMWSA-N
SMILES: CCCCCCC\C=C\C(=O)OCC
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results