accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 736998-56-4

IUPAC Name: 2-[10-(2H-1,3-dithiol-2-ylidene)-3-ethenyl-9,10-dihydroanthracen-9-ylidene]-2H-1,3-dithiole
Synonyms: 2-[10-(1,3-dithiol-2-ylidene)-3-ethenylanthracen-9-ylidene]-1,3-dithiole
Molecular Weight (g/mol): 406.59
Molecular Formula: C22H14S4
InChi Key: PVHQRJPHCJRMKB-UHFFFAOYSA-N
SMILES: C=CC1=CC=C2C(=C1)C(=C1SC=CS1)C1=CC=CC=C1C2=C1SC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results