accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 737000-76-9

IUPAC Name: (3,5-difluoro-2-methoxyphenyl)boronic acid
Synonyms: 3,5-difluoro-2-methoxyphenylboronic acid
Molecular Weight (g/mol): 187.94
Molecular Formula: C7H7BF2O3
InChi Key: JXQHRWOUVJRCSR-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(F)C=C1B(O)O
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results