man

Test

Viewing profile as user:
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Main Menu
Admin
Main Menu
ULS Inventory Manager
Products

CAS RN 737000-77-0

NHFHCl

CAS RN 737000-77-0

IUPAC Name: 3-fluoropiperidine hydrochloride
Synonyms: 3-fluoropiperidine hydrochloride
Molecular Weight (g/mol): 139.6
Molecular Formula: C5H11ClFN
InChi Key: RDJUBLSLAULIAT-UHFFFAOYNA-N
SMILES: Cl.FC1CCCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results