accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73721-78-5

IUPAC Name: 2-acetamido-6-nitrobenzoic acid
Synonyms: 2-acetamido-6-nitrobenzoic acid
Molecular Weight (g/mol): 224.17
Molecular Formula: C9H8N2O5
InChi Key: HHNTZMHFBQIQAK-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(=C1C(O)=O)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results