CAS RN 737776-93-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,1'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ferrocene 98.0+%, TCI America™
CAS: 737776-93-1 Molecular Formula: C22H32B2FeO4 MDL Number: MFCD06411336