accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClClN

CAS RN 7379-35-3

IUPAC Name: hydrogen 4-chloropyridine chloride
Synonyms: hydrogen 4-chloropyridine chloride
Molecular Weight (g/mol): 150
Molecular Formula: C5H5Cl2N
InChi Key: XGAFCCUNHIMIRV-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClC1=CC=NC=C1
Molecular Weight (g/mol) 
  • (8)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (4)
  • (1)
  • (1)
Physical Form 
  • (4)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (5)
  • (8)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)