accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7379-84-2

IUPAC Name: 1,4-bis(2-chloroethyl)benzene
Synonyms: 1,4-bis(2-chloroethyl)benzene
Molecular Weight (g/mol): 203.11
Molecular Formula: C10H12Cl2
InChi Key: GGMHFGIZPKYWGQ-UHFFFAOYSA-N
SMILES: ClCCC1=CC=C(CCCl)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results