accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73819-76-8

IUPAC Name: 5-ethynyl-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 5-ethynyl-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 172.14
Molecular Formula: C10H4O3
InChi Key: DXJLXGJIZZNCBO-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=C1C=CC(=C2)C#C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)