accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOSH

CAS RN 7383-64-4

IUPAC Name: propan-2-yl 3-sulfanylpropanoate
Synonyms: isopropyl 3-sulfanylpropanoate
Molecular Weight (g/mol): 148.22
Molecular Formula: C6H12O2S
InChi Key: XPVDFJPXADVZEN-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)CCS

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results