accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73834-77-2

IUPAC Name: 9H-pyrido[3,4-b]indol-3-amine
Synonyms: 9H-pyrido[3,4-b]indol-3-amine
Molecular Weight (g/mol): 183.21
Molecular Formula: C11H9N3
InChi Key: UGMPOJSWOINMDE-UHFFFAOYSA-N
SMILES: NC1=CC2=C(NC3=C2C=CC=C3)C=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results