accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73842-99-6

CH3CH3CH3SiCH3CH3OHO

CAS RN 73842-99-6

IUPAC Name: 3-[(tert-butyldimethylsilyl)oxy]propan-1-ol
Synonyms: 3-[(tert-butyldimethylsilyl)oxy]propan-1-ol
Molecular Weight (g/mol): 190.36
Molecular Formula: C9H22O2Si
InChi Key: NETUFVYVNJNFMU-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OCCCO
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)