accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73863-44-2

IUPAC Name: 4-chloro-N-(3-methylphenyl)butanamide
Synonyms: 4-chloro-N-(3-methylphenyl)butanamide
Molecular Weight (g/mol): 211.69
Molecular Formula: C11H14ClNO
InChi Key: GBYXVHYDSWAERN-UHFFFAOYSA-N
SMILES: CC1=CC=CC(NC(=O)CCCCl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results