accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHNH2

CAS RN 73874-95-0

IUPAC Name: 4-{[(tert-butoxy)carbonyl]amino}piperidin-1-ium
Synonyms: 4-[(tert-butoxycarbonyl)amino]piperidin-1-ium
Molecular Weight (g/mol): 201.29
Molecular Formula: C10H21N2O2
InChi Key: CKXZPVPIDOJLLM-UHFFFAOYSA-O
SMILES: CC(C)(C)OC(=O)NC1CC[NH2+]CC1
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (2)
  • (5)
  • (2)
Quantity 
  • (2)
  • (3)
  • (4)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (7)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)