accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 7390-81-0

IUPAC Name: 2-hexadecyloxirane
Synonyms: 2-hexadecyloxirane
Molecular Weight (g/mol): 268.49
Molecular Formula: C18H36O
InChi Key: QBJWYMFTMJFGOL-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCCC1CO1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)