accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7391-40-4

IUPAC Name: 2-cyano-N,N-dimethylacetamide
Synonyms: 2-cyano-N,N-dimethylacetamide
Molecular Weight (g/mol): 112.13
Molecular Formula: C5H8N2O
InChi Key: MATJPVGBSAQWAC-UHFFFAOYSA-N
SMILES: CN(C)C(=O)CC#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results