accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73918-56-6

IUPAC Name: 2-(4-bromophenyl)ethan-1-aminium
Synonyms: 2-(4-bromophenyl)ethanaminium
Molecular Weight (g/mol): 201.09
Molecular Formula: C8H11BrN
InChi Key: ZSZCXAOQVBEPME-UHFFFAOYSA-O
SMILES: [NH3+]CCC1=CC=C(Br)C=C1
Boiling Point 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (7)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)