accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7396-58-9

IUPAC Name: bis(decyl)(methyl)amine
Synonyms: bis(decyl)(methyl)amine
Molecular Weight (g/mol): 311.6
Molecular Formula: C21H45N
InChi Key: ATBNMWWDBWBAHM-UHFFFAOYSA-N
SMILES: CCCCCCCCCCN(C)CCCCCCCCCC
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results