accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73960-07-3

IUPAC Name: 4-(difluoromethoxy)benzaldehyde
Synonyms: 4-(difluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: ZWCXOJYJJINQGU-UHFFFAOYSA-N
SMILES: FC(F)OC1=CC=C(C=O)C=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results