accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7399-00-0

IUPAC Name: triethoxy(octadecyl)silane
Synonyms: stearyl triethoxysilane
Molecular Weight (g/mol): 416.76
Molecular Formula: C24H52O3Si
InChi Key: FZMJEGJVKFTGMU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC[Si](OCC)(OCC)OCC
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)