accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOCl

CAS RN 74-11-3

IUPAC Name: 4-chlorobenzoic acid
Synonyms: 4-chlorobenzoic acid chlorodracylic acid acido p-clorobenzoico
Molecular Weight (g/mol): 156.57
Molecular Formula: C7H5ClO2
InChi Key: XRHGYUZYPHTUJZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (2)
  • (5)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (5)
  • (1)