accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3Br

CAS RN 74-83-9

IUPAC Name: bromomethane
Synonyms: methyl bromide methyl bromide, 14C-labeled 14c-labeled methyl bromide detia gas EX-M caswell no. 555 celfume mbc soil fumigant metylu bromek fumigant-1 (obs.) MeBr
Molecular Weight (g/mol): 94.94
Molecular Formula: CH3Br
InChi Key: GZUXJHMPEANEGY-UHFFFAOYSA-N
SMILES: CBr
Solvent or Matrix 
  • (1)
  • (1)
Element 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)