accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3I

CAS RN 74-88-4

IUPAC Name: iodomethane
Synonyms: methyl iodide 14C-labeled methyl iodide 14c-labeled methyl iodide 132I-labeled methyl iodide methyl iodide, 132i-labeled 132i-labeled methyl iodide methyl iodide, 13C-labeled jod-methan 11C-labeled methyl iodide MeI
Molecular Weight (g/mol): 141.94
Molecular Formula: CH3I
InChi Key: INQOMBQAUSQDDS-UHFFFAOYSA-N
SMILES: CI
Molecular Weight (g/mol) 
  • (26)
  • (12)
Quantity 
  • (1)
  • (1)
  • (1)
  • (7)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (17)
  • (3)
  • (2)
  • (8)
  • (4)
Percent Purity 
  • (1)
  • (1)
  • (4)
  • (10)
Physical Form 
  • (16)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (12)
  • (4)
  • (1)
  • (17)
  • (26)
  • (12)
  • (1)
  • (1)
  • (1)
  • (7)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (17)
  • (3)
  • (2)
  • (8)
  • (4)
  • (1)
  • (1)
  • (4)
  • (10)
  • (16)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)