accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74-94-2

IUPAC Name: dimethylamine borane
Synonyms: dimethylamine borane
Molecular Weight (g/mol): 58.92
Molecular Formula: C2H10BN
InChi Key: FTDUHBOCJSQEKS-UHFFFAOYSA-N
SMILES: B.CNC
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (4)
  • (4)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (4)
Physical Form 
  • (2)
  • (3)
  • (4)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (6)
  • (9)
  • (3)
  • (4)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (3)
  • (4)
  • (2)