accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7400-45-5

IUPAC Name: methyl 3-[(2-methoxy-2-oxoethyl)sulfanyl]propanoate
Synonyms: methyl 3-[(2-methoxy-2-oxoethyl)sulfanyl]propanoate
Molecular Weight (g/mol): 192.23
Molecular Formula: C7H12O4S
InChi Key: XNDKLLFGXIEGKL-UHFFFAOYSA-N
SMILES: COC(=O)CCSCC(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results