accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74011-58-8

IUPAC Name: 1-ethyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid
Synonyms: enoxacin faulding brand of enoxacin ENX sesquihydrate, enoxacin pierre fabre brand of enoxacin sesquihydrate enoxacin preparation comprecin enoxacin (substance) flumark enoxacin sesquihydrate
Molecular Weight (g/mol): 320.32
Molecular Formula: C15H17FN4O3
InChi Key: IDYZIJYBMGIQMJ-UHFFFAOYSA-N
SMILES: CCN1C=C(C(O)=O)C(=O)C2=CC(F)=C(N=C12)N1CCNCC1
Color 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Type 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results