accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7411-49-6

IUPAC Name: tetrahydrogen [1,1'-biphenyl]-3,3',4,4'-tetramine tetrachloride
Synonyms: tetrahydrogen 3,3'-diaminobenzidine tetrachloride
Molecular Weight (g/mol): 360.1
Molecular Formula: C12H18Cl4N4
InChi Key: KJDSORYAHBAGPP-UHFFFAOYSA-N
SMILES: [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NC1=CC=C(C=C1N)C1=CC=C(N)C(N)=C1
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)