accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CNBrBr

CAS RN 741293-42-5

IUPAC Name: 3,6-dibromo-9-(4-tert-butylphenyl)-9H-carbazole
Synonyms: 3,6-dibromo-9-(4-tert-butylphenyl)carbazole
Molecular Weight (g/mol): 457.21
Molecular Formula: C22H19Br2N
InChi Key: JIWBCHGVZITSFJ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)N1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results