accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74137-36-3

IUPAC Name: 1,3-dibromo-5-methoxybenzene
Synonyms: 1,3-dibromo-5-methoxybenzene
Molecular Weight (g/mol): 265.93
Molecular Formula: C7H6Br2O
InChi Key: OQZAQBGJENJMHT-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=CC(Br)=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)